1,2-Bis(diphenylphosphino)benzene ≥98%

Supplier: ACROS ORGANICS MS

AC36375-0010
200021-566EA 90.52 USD
200021-566
1,2-Bis(diphenylphosphino)benzene ≥98%
1,2-Bis(diphenylphosphino)benzene
Formula: C₃₀H₂₄P₂
Storage Temperature: Ambient
MDL Number: MFCD00014081
CAS Number: 13991-08-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 184°C to 189°C
Assay ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR