N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide 98%

Warning

Synonyms: (1R,2R)-(-)-N-(4-toluenesulfonyl)-1,2-diphenylethylenediamine(1R,2R)-(-)-N-(4-Toluenesulphonyl)-1,2-diphenylethylenediamine(1R,2R)-(-)-N-(4-Toluenesulfonyl)-1,2-diphenylethylenediamine(R,R)-TsDPEN(1R,2R)-(-)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine

AC36368-5000
200051-188EA 70.5 USD
200051-188
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide 98%
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide

Appearance: White to light yellow Crystalline powder or crystals

Formula: C₂₁H₂₂N₂O₂S
MW: 366.48 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD02093428
CAS Number: 144222-34-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 126 to 130 °C
Specific optical rotation -57.5° to -62.5° (20 °C, 589 nm) (c=1, CH2Cl2)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR