(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%, 98% ee
AC36299-0010
AC36299-0050
200015-316EA
828
USD
200015-316
200008-224
(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%, 98% ee
(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride
Appearance: White Powder
Formula:
C₁₁H₁₆ClNO₂ Melting Pt: 119 °C Storage Temperature: Ambient |
CAS Number:
340188-50-3 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Enantiomeric excess | ≥97.5 % (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...