(R)-3-(Boc-amino)-3-phenylpropionic acid ≥95%, ee 98%

Synonyms: (R)-N-BOC-3-Amino-3-phenylpropanoic acid(R)-3-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid

AC36286-5000 AC36286-2500 AC36286-0010
200027-368EA 300.5 USD
200027-368 200039-494 103512-718
(R)-3-(Boc-amino)-3-phenylpropionic acid ≥95%, ee 98%
(R)-3-(Boc-amino)-3-phenylpropionic acid
Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320859
CAS Number: 161024-80-2

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR