Boc-D-propargylglycine 95%, 98% ee

Warning

Synonyms: Boc-D-Pra-OHN-Boc-2-propargyl-D-glycine(R)-2-((tert-Butoxycarbonyl)amino)pent-4-ynoic acid(R)-N-BOC-Propargylglycine

AC36285-0050 AC36285-0010
200015-326EA 366 USD
200015-326 103512-716
Boc-D-propargylglycine 95%, 98% ee
Boc-D-propargylglycine
Formula: C₁₀H₁₅NO₄
MW: 213.233 g/mol
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD01320886
CAS Number: 63039-46-3

Order Now

Specification Test Results

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR