(1R,4S)-4-Aminocyclopent-2-enecarboxylic acid 95%, 98% ee

Warning

AC36226-0010
200004-512EA 284.9 USD
200004-512
(1R,4S)-4-Aminocyclopent-2-enecarboxylic acid 95%, 98% ee
(1R,4S)-4-Aminocyclopent-2-enecarboxylic acid

Appearance: Brown Crystalline powder or flakes

Formula: C₆H₉NO₂
Melting Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00211285
CAS Number: 134003-04-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown Crystalline powder or flakes
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 % (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR