(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one 95%, 98% ee

AC36222-2500
200027-262EA 156.4 USD
200027-262
(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one 95%, 98% ee
(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one

Appearance: White to light yellow Crystalline powder

Formula: C₆H₉NO
Melting Pt: 76.2 °C
Storage Temperature: Ambient
CAS Number: 134003-03-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR