5-Aminonicotinic acid 95%

Warning

Synonyms: 5-Amino-3-pyridinecarboxylic acid

AC36114-0050
103532-592EA 352 USD
103532-592
5-Aminonicotinic acid 95%
5-Aminonicotinic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129116
CAS Number: 24242-19-1
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Golden to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR