Silica gel C8-RP 12%C, ∼ 1.2 mmol/g 40-63 µm

AC36014-2500 AC36014-0500
103513-310EA 1087.69 USD
103513-310 200056-290
Silica gel C8-RP 12%C, ∼ 1.2 mmol/g 40-63 µm
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size 40 to 63 µm
Elemental Analysis ≥11.6 % (C)
Loading ≥1.21 mmol/g
Volatile matter ≤4 % (110 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR