Isobutylboronic acid 98%
|
Warning
|
Synonyms:
(2-Methylpropyl)boronic acid
AC35995-0050
AC35995-0010
200015-248EA
25.1
USD
200015-248
200004-518
Isobutylboronic acid 98%
Isobutylboronic acid
Formula:
C₄H₁₁BO₂ MW: 101.94 g/mol Melting Pt: 108…111 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00134156 CAS Number: 84110-40-7 |
Specification Test Results
Appearance | White to cream Crystalline powder or flakes |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...