3-Amino-6-chloro-4-methylpyridine 98%

Warning

Synonyms: 6-Chloro-4-methylpyridin-3-amine5-Amino-2-chloro-4-methylpyridine3-Amino-6-chloro-4-picoline

AC36099-0050 AC36099-0010
103532-582EA 360 USD
103532-582 200054-368
3-Amino-6-chloro-4-methylpyridine 98%
3-Amino-6-chloro-4-methylpyridine
Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD03427661
CAS Number: 66909-38-4
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Bordeaux to brown Powder
Infrared spectrum Conforms
Melting point 69 to 73 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR