2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Warning

Synonyms: Isopropyl pinacolyl borate4,4,5,5-tetramethyl-2-(propan-2-yloxy)-1,3,2-dioxaborolane

AC35949-0010
200028-812EA 15.1 USD
200028-812
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₉H₁₉BO₃
MW: 186.06 g/mol
Boiling Pt: 73 °C (15 mmHg)
Density: 0.912 g/cm³
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192241
CAS Number: 61676-62-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4100 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR