4-(N,N-Dimethylamino)phenylboronic acid 96%

Warning

Synonyms: 4-(Dimethylamino)benzeneboronic acid

AC35923-5000 AC35923-0025
200017-260EA 42.7 USD
200017-260 103515-244
4-(N,N-Dimethylamino)phenylboronic acid 96%
4-(N,N-Dimethylamino)phenylboronic acid
Formula: C₈H₁₂BNO₂
MW: 165.00
Melting Pt: 227 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074642
CAS Number: 28611-39-4

Order Now

Specification Test Results

Appearance White to grey Powder and/or lumps
Infrared spectrum Conforms
Titration with NaOH ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR