4-(Aminomethyl)phenylboronic acid hydrochloride 96%

Warning

Synonyms: 4-Aminomethylphenylboronic acid hydrochloride4-(Aminomethyl)benzeneboronic acid hydrochloride

AC35884-2500
200005-368EA 124 USD
200005-368
4-(Aminomethyl)phenylboronic acid hydrochloride 96%
4-(Aminomethyl)phenylboronic acid hydrochloride
Formula: C₇H₁₁BClNO₂
MW: 187.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01632199
CAS Number: 75705-21-4

Order Now

Specification Test Results

Appearance Light yellow Powder
Infrared spectrum Conforms
Titration Argentometric ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR