Nickel(II) tetrafluoroborate hexahydrate 99%

Danger

AC35873-0100
200044-210EA 43.8 USD
200044-210
Nickel(II) tetrafluoroborate hexahydrate 99%
Nickel (II) tetrafluoroborate hexahydrate
Formula: B₂F₈Ni·6H₂O
Storage Temperature: Ambient
MDL Number: MFCD00016249
CAS Number: 15684-36-3
EINECS: 238-753-4
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance Blue-green Powder or crystals
Titration Complexometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR