Lithium trifluoromethanesulfonate 98%

Warning

AC35871-1000 AC35871-0250 AC35871-0050
200025-700EA 24.3 USD
200025-700 200025-698 103515-212
Lithium trifluoromethanesulfonate 98%
Lithium trifluoromethanesulphonate
Formula: CF₃LiO₃S
Melting Pt: 423 °C
Storage Temperature: Ambient
MDL Number: MFCD00013227
CAS Number: 33454-82-9
EINECS: 251-528-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Assay 4.34 to 4.56 % (Li)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR