Benzothiazol-2-ylmethanol 98%

Warning

Synonyms: 2-(Hydroxymethyl)benzothiazole2-Benzothiazolemethanol2-Hydroxymethylbenzothiazole

AC35850-0010
200005-074EA 41.2 USD
200005-074
Benzothiazol-2-ylmethanol 98%
Benzothiazol-2-ylmethanol
Formula: C₈H₇NOS
MW: 165.21 g/mol
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00226293
CAS Number: 37859-42-0

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 98 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR