Dipentyl phthalate 97%

Danger

Synonyms: Di-n-pentyl phthalate

AC35736-5001 AC35736-1000 AC35736-0250
103527-820EA 1779 USD
103527-820 200012-134 200006-354
Dipentyl phthalate 97%
Dipentyl phthalate
Formula: C₁₈H₂₆O₄
MW: 306.41 g/mol
Boiling Pt: 342 °C (760 mmHg)
Melting Pt: –55 °C
Density: 1.023 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00041934
CAS Number: 131-18-0
EINECS: 205-017-9
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4885 to 1.4905 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR