1-Methyl-3-octylimidazolium hexafluorophosphate 98+%
|
Warning
|
Synonyms:
1-Octyl-3-methylimidazolium hexafluorophosphate
AC35676-0050
AC35676-0250
200063-756EA
60.3
USD
200063-756
200034-002
1-Methyl-3-octylimidazolium hexafluorophosphate 98+%
1-Methyl-3-octylimidazolium hexafluorophosphate
Formula:
C₁₂H₂₃F₆N₂P MW: 340.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06798203 CAS Number: 304680-36-2 |
Specification Test Results
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4230 to 1.4250 (20 °C, 589 nm) |
Water | ≤1.0 % (K.F.) |
NMR (1H-NMR) | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...