Phorbol-12-myristate-13-acetate 97%

Warning

Synonyms: TPA12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate

AC35615-0010 AC35615-0050
103513-246EA 219 USD
103513-246 200003-528
Phorbol-12-myristate-13-acetate 97%
Phorbol-12-myristate-13-acetate
Formula: C₃₆H₅₆O₈
MW: 616.836 g/mol
Melting Pt: 72 °C
Storage Temperature: Freezer
MDL Number: MFCD00036736
CAS Number: 16561-29-8

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Film or foam
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR