Methyl-5-bromo-2-furoate 98%

Warning

Synonyms: Methyl 5-bromofuran-2-carboxylateMethyl 5-Bromo-2-furancarboxylate5-Bromo-2-furoic acid methyll ester5-Bromo-2-furancarboxylic acid methyll esterMethyl 5-Bromo-2-furoate

AC35575-0050 AC35575-0250
103513-244EA 646 USD
103513-244 103513-242
Methyl-5-bromo-2-furoate 98%
Methyl 5-bromo-2-furoate
Formula: C₆H₅BrO₃
MW: 205.01 g/mol
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00092313
CAS Number: 2527-99-3

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 63 to 68 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR