2-(Chloromethyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole ≥98%

Warning

AC35471-0010
200022-576EA 102.3 USD
200022-576
2-(Chloromethyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole ≥98%
2-(Chloromethyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole

Appearance: Light yellow Crystalline powder

Formula: C₁₀H₉ClN₂O₂
Melting Pt: 91…95 °C
Storage Temperature: Ambient
CAS Number: 24023-71-0

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 91°C to 95°C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR