TentaGel® MB-Br

AC35448-0010
200056-430EA 77.8 USD
200056-430
TentaGel® MB-Br
TentaGel® MB-Br
Storage Temperature: Refrigerator
MDL Number: MFCD00217898
CAS Number: 73342-17-3

Order Now

Specification Test Results

Appearance White to yellow Powder
Particle size 140 to 170 µm
Loading 0.4 to 0.6 mmol/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR