2-Acetamido-5-bromobenzoic acid ≥97%

AC35402-0050
200028-438EA 28.9 USD
200028-438
2-Acetamido-5-bromobenzoic acid ≥97%
2-Acetamido-5-bromobenzoic acid
Formula: C₉H₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD00040904
CAS Number: 38985-79-4

Order Now

Specification Test Results

Appearance Light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 218 to 223 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR