Morin hydrate

Synonyms: 2',3,4',5,7-Pentahydroxyflavone

AC35400-0100
200025-952EA 72.7 USD
200025-952
Morin hydrate
Morin hydrate
Formula: C₁₅H₁₀O₇
MW: 302.24 g/mol
Melting Pt: 300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00217054
CAS Number: 654055-01-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
UV spectrum Complies
Water ≤7 %
Sulfated ash ≤2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR