Cyanogen bromide 3 M in dichloromethane

Danger

AC35399-1000
200057-390EA 117.69 USD
200057-390
Cyanogen bromide 3 M in dichloromethane
Cyanogen bromide
Formula: CBrN
MW: 105.92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011597
CAS Number: 506-68-3
EINECS: 208-051-2
UN: 2927
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Strength 2.95 to 3.3 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR