Triacetyllevoglucosan 98+%
Synonyms:
1,6-Anhydro-β-D-glucose-2,3,4-tri-O-acetate
AC35368-5000
200051-468EA
578.22
USD
200051-468
Triacetyllevoglucosan 98+%
Triacetyllevoglucosan
Appearance: White to off-white Crystalline powder
Formula:
C₁₂H₁₆O₈ MW: 288.252 g/mol Melting Pt: 111…112 °C Storage Temperature: Ambient |
CAS Number:
13242-55-2 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
TLC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...