3,5-Dimethyl-4-hydroxybenzonitrile 97%
|
Warning
|
Synonyms:
4-Hydroxy-3,5-dimethylbenzonitrile
AC35366-0050
200005-190EA
107
USD
200005-190
3,5-Dimethyl-4-hydroxybenzonitrile 97%
3,5-Dimethyl-4-hydroxybenzonitrile
Formula:
C₉H₉NO MW: 147.18 g/mol Melting Pt: 123…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00221716 CAS Number: 4198-90-7 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 123 to 126 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...