3'-Benzyloxyacetophenone 97%

Warning

Synonyms: 1-(3-(Benzyloxy)phenyl)ethanone1-[3-(Benzyloxy)phenyl]ethan-1-one

AC35312-0050
200023-176EA 32.5 USD
200023-176
3'-Benzyloxyacetophenone 97%
3'-Benzyloxyacetophenone
Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026221
CAS Number: 34068-01-4
EINECS: 251-816-0

Order Now

Specification Test Results

Appearance Light yellow Low melting solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR