Pyridoxal hydrochloride 99%

AC35271-0250 AC35271-0050 AC35271-1000 AC35271-0010 AC35271-5000
103466-422EA 2224 USD
103466-422 103466-420 200038-682 200020-556 200020-558
Pyridoxal hydrochloride 99%
Pyridoxal hydrochloride
Formula: C₈H₁₀ClNO₃
Melting Pt: 171…175 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012809
CAS Number: 65-22-5
EINECS: 200-602-5

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 171 to 175 °C (Dec)
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR