(R)-(-)-Tetrahydrofurfurylamine 98+%
|
Warning
|
Synonyms:
(R)-2-Tetrahydrofurfurylamine
, (R)-(-)-2-(Aminomethyl)tetrahydrofuran
AC35263-0010
200003-506EA
84.5
USD
200003-506
(R)-(-)-Tetrahydrofurfurylamine 98+%
(R)-(-)-Tetrahydrofurfurylamine
Appearance: Clear colorless Liquid
Formula:
C₅H₁₁NO MW: 101.15 g/mol Boiling Pt: 151…152 °C (760 mmHg) Density: 0.98 g/cm³ Flash Pt: 46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192476 CAS Number: 7202-43-9 UN: 2943 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4540 to 1.4560 (20 °C, 589 nm) |
GC | ≥98.0 % |
Specific optical rotation | -11° to -16° (20 °C, 589 nm) (c=4, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...