Isosorbide 98%

Synonyms: D-Isosorbide(3R,3aR,6S,6aR)-Hexahydrofuro[3,2-b]furan-3,6-diol

AC35191-0050 AC35191-1000
200006-744EA 68.5 USD
200006-744 200043-718
Isosorbide 98%
Isosorbide
Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Melting Pt: 56…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00064827
CAS Number: 652-67-5

Order Now

Specification Test Results

Appearance Off-white to yellow to beige Crystals or crystalline solid or mass
Infrared spectrum Conforms
Melting point 56 to 62 °C
GC ≥97.5 %
Specific optical rotation +42° to +48° (20 °C, 589 nm) (c=3, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR