trans-4-Methoxycinnamaldehyde 98%
|
Warning
|
Synonyms:
(E)-3-(4-Methoxyphenyl)acrylaldehyde
AC35171-0250
AC35171-1000
AC35171-0050
200027-038EA
55
USD
200027-038
200014-908
103514-020
trans-4-Methoxycinnamaldehyde 98%
trans-4-Methoxycinnamaldehyde
Formula:
C₁₀H₁₀O₂ MW: 162.19 Boiling Pt: 145 °C Melting Pt: 55…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017343 CAS Number: 24680-50-0 |
Specification Test Results
Appearance | Yellow to light orange Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 55 to 60 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...