Sodium glycolate 97%

AC35157-0250 AC35157-1000
200032-822EA 47.6 USD
200032-822 200014-486
Sodium glycolate 97%
Sodium glycolate
Formula: C₂H₃NaO₃
MW: 98.03 g/mol
Storage Temperature: Ambient
EINECS: 220-624-9

Order Now

Specification Test Results

Appearance White to cream Powder and/or lumps
Infrared spectrum Conforms
Titration with HClO4 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR