2-Naphthalenemethanol 98%

AC35115-0050 AC35115-0250 AC35115-0010
200054-200EA 94.6 USD
200054-200 200054-198 103463-576
2-Naphthalenemethanol 98%
2-Naphthalenemethanol
Formula: C₁₁H₁₀O
MW: 158.2000 g/mol
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00004124
CAS Number: 1592-38-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR