2-Methylphenethylamine 97%
|
Danger
|
Synonyms:
2-(o-Tolyl)ethylamine
AC35110-0050
AC35110-0010
200009-526EA
46.8
USD
200009-526
200009-528
2-Methylphenethylamine 97%
2-Methylphenethylamine
Formula:
C₉H₁₃N Storage Temperature: Ambient |
MDL Number:
MFCD01310827 CAS Number: 55755-16-3 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5340 to 1.5360 (20 °C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...