3-Fluorophenethylamine 99%

Danger

Synonyms: 2-(3-Fluorophenyl)ethylamine

AC35094-0250 AC35094-1000 AC35094-0050
200009-930EA 76.8 USD
200009-930 200009-928 103530-168
3-Fluorophenethylamine 99%
3-Fluorophenethylamine
Formula: C₈H₁₀FN
Boiling Pt: 87 °C (15 mmHg)
Density: 1.066 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00075376
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5080 to 1.5100 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR