Diphenhydramine hydrochloride 99%

Warning

Synonyms: 2-(Benzhydryloxy)-N,N-dimethylethylamine Hydrochloride2-Diphenylmethoxy-N,N-dimethylethylamine Hydrochloride2-(Benzhydryloxy)-N,N-dimethylethanamine hydrochloride

AC35085-1000 AC35085-0250
200012-136EA 42.7 USD
200012-136 200043-314
Diphenhydramine hydrochloride 99%
Diphenhydramine hydrochloride

Appearance: White Adhering crystalline powder or crystals

Formula: C₁₇H₂₁NO
MW: 291.82 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00012479
CAS Number: 147-24-0

Order Now

Specification Test Results

Appearance White Adhering crystalline powder or crystals
Infrared spectrum Conforms
Melting point 168 to 172 °C
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR