3,4-Dichlorophenethylamine 99%

Warning

AC35080-0250 AC35080-0050
103530-166EA 836 USD
103530-166 200003-484
3,4-Dichlorophenethylamine 99%
3,4-Dichlorophenethylamine
Formula: C₈H₉Cl₂N
Boiling Pt: 280 °C
Density: 1.268 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00060617
CAS Number: 21581-45-3
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5655 to 1.5675 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR