2,4-Dichlorophenethylamine 98%

Warning

AC35078-0250 AC35078-0050 AC35078-1000
103514-002EA 364.3 USD
103514-002 200022-180 200016-094
2,4-Dichlorophenethylamine 98%
2,4-Dichlorophenethylamine

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5645 to 1.5665 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR