Pentaethylene glycol 98%

Warning

Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diol

AC35073-0050 AC35073-0250
200013-920EA 269 USD
200013-920 200020-320
Pentaethylene glycol 98%
Pentaethylene glycol
Formula: C₁₀H₂₂O₆
MW: 238.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002878
CAS Number: 4792-15-8
EINECS: 225-341-4

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR