(3-Bromophenyl)acetone 98+%
AC35060-0250
AC35060-0050
103463-670EA
437
USD
103463-670
200003-478
(3-Bromophenyl)acetone 98+%
(3-Bromophenyl)acetone
Specification Test Results
Appearance | Clear light yellow to dark yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5570 to 1.5600 (20 °C, 589 nm) |
GC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...