(2-Bromophenyl)acetone 99%
Synonyms:
1-(2-Bromophenyl)propan-2-one
AC35059-0050
AC35059-0010
103464-406EA
36.6
USD
103464-406
200009-246
(2-Bromophenyl)acetone 99%
(2-Bromophenyl)acetone
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5575 to 1.5595 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...