Chloro(diethyl)phosphine 95%

Danger

Synonyms: Diethylchlorophosphine

AC35031-0050 AC35031-0010
103527-792EA 239 USD
103527-792 200036-798
Chloro(diethyl)phosphine 95%
Chloro(diethyl)phosphine
Formula: C₄H₁₀ClP
MW: 124.55 g/mol
Boiling Pt: 131…132 °C (760 mmHg)
Density: 1.023 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00013621
CAS Number: 686-69-1
EINECS: 211-689-4
UN: 3129
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20 °C, 589 nm)
NMR ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR