D-ɑ-Tocopherylquinone 90-95%
Synonyms:
Vitamin E Quinone (D-form)
AC35030-5000
200018-690EA
419
USD
200018-690
D-ɑ-Tocopherylquinone 90-95%
D-alpha-Tocopherylquinone
Appearance: Clear amber to red Viscous liquid
Formula:
C₂₉H₅₀O₃ MW: 446.72 g/mol Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00135657 CAS Number: 7559-04-8 |
Specification Test Results
Appearance | Clear amber to red Viscous liquid |
Infrared spectrum | Conforms |
GC | ≥90 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...