1H,1H,2H,2H-Heptadecafluorodecyl acrylate 97% stabilized

Warning

Synonyms: Acrylic Acid 1H,1H,2H,2H-Heptadecafluorodecyl Ester

AC34861-0050
200033-946EA 51.2 USD
200033-946
1H,1H,2H,2H-Heptadecafluorodecyl acrylate 97% stabilized
1H,1H,2H,2H-Heptadecafluorodecyl acrylate

Appearance: Clear colorless to light yellow Liquid

Formula: C₁₃H₇F₁₇O₂
MW: 518.17 g/mol
Boiling Pt: 100 °C (4 mmHg)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042306
CAS Number: 27905-45-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3360 to 1.3380 (20 °C, 589 nm)
GC ≥96.0 %
Stabilizer Approx. 15 ppm TBC and 70 ppm BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR