Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane) 98%

Danger

Synonyms: Isobutylene oxide

AC34850-0250 AC34850-1000
200012-830EA 236 USD
200012-830 200012-832
Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane) 98%
Isobutylene oxide (2,2-Dimethyloxirane, 1,2-Epoxy-2-methylpropane)
Formula: C₄H₈O
Flash Pt: –32 °C
Storage Temperature: Ambient
MDL Number: MFCD00066354
CAS Number: 558-30-5
EINECS: 209-193-8
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3705 to 1.3745 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR