(3R,4R)-1-Benzylpyrrolidine-3,4-diol 97%

Warning

Synonyms: (3R,4R)-(-)-1-Benzyl-3,4-pyrrolidindiol

AC34872-0010 AC34872-0050
200008-172EA 175 USD
200008-172 103513-934
(3R,4R)-1-Benzylpyrrolidine-3,4-diol 97%
(3R,4R)-1-Benzylpyrrolidine-3,4-diol
Formula: C₁₁H₁₅NO₂
MW: 193.2423 g/mol
Melting Pt: 95…101 °C
Storage Temperature: Ambient
MDL Number: MFCD01073894
CAS Number: 163439-82-5

Order Now

Specification Test Results

Appearance Off-white to yellow to light brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 95 to 101 °C
GC ≥96.0 %
Specific optical rotation -29° to -35° (20 °C, 589 nm) (c=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR