(S)-(-)-2-(Diphenylmethyl)pyrrolidine 97+%

Warning

Synonyms: (S)-2-Diphenylmethylpyrrolidine

AC34837-0010 AC34837-2500 AC34837-0050
200021-406EA 639 USD
200021-406 200021-404 103513-930
(S)-(-)-2-(Diphenylmethyl)pyrrolidine 97+%
(S)-(-)-2-(Diphenylmethyl)pyrrolidine
Formula: C₁₇H₁₉N
MW: 237.35 g/mol
Density: 1.062 g/cm³
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00799525
CAS Number: 119237-64-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5860 to 1.5880 (20 °C, 589 nm)
GC ≥97.0 %
Specific optical rotation -2° to -4° (25 °C, 589 nm) (c=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR