Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methylmorpholine-N-oxide 50% (w/w) in water

Supplier: ACROS ORGANICS MS
Warning
Synonyms: 4-Methylmorpholine N-Oxide

AC34827-0010 AC34827-0250 AC34827-1000
102775-598 102773-850 102774-810
N-Methylmorpholine-N-oxide 50% (w/w) in water


Formula: C₅H₁₁NO₂
MW: 117,15 g/mol
Density: 1,13 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00005947
CAS Number: 7529-22-8
EINECS: 231-391-8
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Solution
Strength 49 to 51 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty